VWR®, Macro Pipette Tips
Macro pipette tips, 1000 - 5000 µl, +, +, 123,1 mm, Clear, Eppendorf, Socorex, Sartorius Biohit, Rack of 50 tips
Nome del fornitore: VWR Collection
Vai a
Product Details & Documents
Improved accuracy over serological pipettes.
- Tip length facilitates access into bottles, cell culture flasks, and tubes
Universal fit for compatibility with most major pipettor brands.
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.
Specifiche
- Confezione:Rack of 50 tips
- Sterili:+
- Volume:1000 - 5000 µl
- Lunghezza:123,1 mm
- Seq:02
- Stile punta:Macro pipette tips
- Colore puntale:Clear
- Per uso con:Eppendorf, Socorex, Sartorius Biohit
- Filtro:+