[EN]SOFT CARE SENSITIVE 6X1L W2692 1 * 6 Bott.
DIVY101108190
: Diversey
Frequently Bought Together
2-Propanolo ≥99.0% USP, Ph. Eur.
: VWR Chemicals
ter-Butil metil etere, extra dry over Molecular Sieve 99+%, AcroSeal®
: Thermo Fisher Scientific
N,N-Dimetilformammide 99+%, extra puro
: Thermo Fisher Scientific
Acqua, extra puro, deionised
: Thermo Fisher Scientific
1-Clorobutano 99.5% per analisi
: Thermo Fisher Scientific
TOLUENE (REAG. PH. EUR.) FOR ANALYSIS, A 1 * 2,5 L
: PanReac AppliChem
Tetraidrofurano 99.8% non stabilizzato per HPLC
: Thermo Fisher Scientific
1-Butanolo ≥99.7%, CHROMASOLV™ Plus per HPLC, Riedel-de Haën™
: Honeywell Chemicals
Acetone 99.8% per HPLC
: Thermo Fisher Scientific
Acetone, extra dry 99.8%, AcroSeal®
: Thermo Fisher Scientific
Esano (miscela di isomeri) ≥95%, Laboratory Reagent
: Honeywell Chemicals