Lid 201 - 454x320x20 PS-FS Natural Zubehör zu #216-1763, Lagerbehälter 23 L 1 * 25 ST
Catalog # LINP9322000000
Supplier: LINPAC
Lid 201 - 454x320x20 PS-FS Natural Zubehör zu #216-1763, Lagerbehälter 23 L 1 * 25 ST
Catalog # LINP9322000000
Supplier: LINPAC
Frequently Bought Together
Masterflex® I/P® Precision Pump Tubing, Versilon™ A-60-G, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: Avantor Fluid Handling
BAC40-001-TFF1-II-02-0 1 * 1 ST
Supplier: JM SEPARATIONS
Tubi, gomma
Supplier: KARTELL
Catalyst Pump Tubing Links / Assemblies for FH15, FH30 and FH100M Pumps
Supplier: Avantor Fluid Handling
Bev-A-Line® V Transfer Pump Tubing
Supplier: Avantor Fluid Handling
[EN]TYGON LFL 1/8'' X 1/4'' 25'/7.62M 1 * 25 Piedi
Supplier: Avantor Fluid Handling
[EN]ULTRA PLAT SIL 3/8X7/16 50FT 1 * 1 pezzi
Supplier: Avantor Fluid Handling
[EN]MT-1886/1861 - 2 L GLASS FLASK 1 * 5 pezzi
Supplier: JM SEPARATIONS
NEOTERYX 10UL MITRA AUTORACK (96-SAMPLER) : NO BARCODE
Supplier: MARKET SOURCE PART PROCESS
Samplers, PowderThief
Supplier: Sampling Systems
Two-dimensional BioProcess Container™ system, Labtainer™ BPC bags
Supplier: THERMO BIOPROCESS DIVISION