Orders LinkContactUsLinkComponent
Electronic Battery Crimping Tools
Electronic Battery Crimping Tools
Electronic flip off crimping tool for 20 mm flip off crimp caps
Codice catalogo SGEA6A20F0
Nome del fornitore:   Trajan Scientific and Medical
Electronic Battery Crimping Tools
Codice catalogo SGEA6A20F0
Nome del fornitore:  Trajan Scientific and Medical
Electronic Battery Crimping Tools
Electronic Battery Crimping Tools
Electronic flip off crimping tool for 20 mm flip off crimp caps
Price
SFr. 1’130.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Electronic battery crimping tools effortlessly crimp/decap aluminum and 2-part aluminum/steel caps with just the push of a button.
  • Consistently crimp caps to a reproducible seal setting with less physical strain
  • Powerful brushless motor with a fast cycle
  • Long lasting lithium ion battery provides up to 800 crimps/decaps per charge with an average recharge time of 1 to 2 hours
  • Innovative charging system, which includes a warning light when recharging is necessary
  • Can be operated while charging
  • The controls to increase or decrease crimp settlings are conveniently located on top of the unit, preventing accidental adjustment of the settings
  • OLED screen for simple operation displays tool size, battery status, crimp setting and cycle result
  • Performance tracking through an internal cycle log
Tools in 8, 11, 13, and 20 mm crimping and decapping.
Specifiche
  • Descrizione:
    Electronic flip off crimping tool for 20 mm flip off crimp caps
  • Seq:
    06
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Stand for CRS electronic and hand crimpers and decappers
    Stand for CRS electronic and hand crimpers and decappers
    SGEA5ASTA0
    Log in per disponibilità
    1 * 1 ST - SFr. 84.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Base for electronic crimping tool and high powered crimping tool
    Base for electronic crimping tool and high powered crimping tool
    SGEA5ABAS0
    Log in per disponibilità
    1 * 1 ST - SFr. 278.00