Orders LinkContactUsLinkComponent
Electronic Battery Crimping Tools
Electronic Battery Crimping Tools
Electronic decapper tool for 20 mm aluminum crimp caps
Codice catalogo SGEA6A20D0
Nome del fornitore:   Trajan Scientific and Medical
Electronic Battery Crimping Tools
Codice catalogo SGEA6A20D0
Nome del fornitore:  Trajan Scientific and Medical
Electronic Battery Crimping Tools
Electronic Battery Crimping Tools
Electronic decapper tool for 20 mm aluminum crimp caps
Price
SFr. 1’130.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Electronic battery crimping tools effortlessly crimp/decap aluminum and 2-part aluminum/steel caps with just the push of a button.
  • Consistently crimp caps to a reproducible seal setting with less physical strain
  • Powerful brushless motor with a fast cycle
  • Long lasting lithium ion battery provides up to 800 crimps/decaps per charge with an average recharge time of 1 to 2 hours
  • Innovative charging system, which includes a warning light when recharging is necessary
  • Can be operated while charging
  • The controls to increase or decrease crimp settlings are conveniently located on top of the unit, preventing accidental adjustment of the settings
  • OLED screen for simple operation displays tool size, battery status, crimp setting and cycle result
  • Performance tracking through an internal cycle log
Tools in 8, 11, 13, and 20 mm crimping and decapping.
Specifiche
  • Descrizione:
    Electronic decapper tool for 20 mm aluminum crimp caps
  • Seq:
    09
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Stand for CRS electronic and hand crimpers and decappers
    Stand for CRS electronic and hand crimpers and decappers
    SGEA5ASTA0
    Log in per disponibilità
    1 * 1 ST - SFr. 84.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Base for electronic crimping tool and high powered crimping tool
    Base for electronic crimping tool and high powered crimping tool
    SGEA5ABAS0
    Log in per disponibilità
    1 * 1 ST - SFr. 278.00