Electronic Battery Crimping Tools
Electronic crimper tool for 20 mm crimp caps
Codice catalogo SGEA6A20C0
Nome del fornitore: Trajan Scientific and Medical
Electronic Battery Crimping Tools
Codice catalogo SGEA6A20C0
Nome del fornitore: Trajan Scientific and Medical
Vai a
Product Details & Documents
Electronic battery crimping tools effortlessly crimp/decap aluminum and 2-part aluminum/steel caps with just the push of a button.
- Consistently crimp caps to a reproducible seal setting with less physical strain
- Powerful brushless motor with a fast cycle
- Long lasting lithium ion battery provides up to 800 crimps/decaps per charge with an average recharge time of 1 to 2 hours
- Innovative charging system, which includes a warning light when recharging is necessary
- Can be operated while charging
- The controls to increase or decrease crimp settlings are conveniently located on top of the unit, preventing accidental adjustment of the settings
- OLED screen for simple operation displays tool size, battery status, crimp setting and cycle result
- Performance tracking through an internal cycle log
Tools in 8, 11, 13, and 20 mm crimping and decapping.
Specifiche
- Descrizione:Electronic crimper tool for 20 mm crimp caps
- Seq:05
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Stand for CRS electronic and hand crimpers and decappersStand for CRS electronic and hand crimpers and decappersSGEA5ASTA0Log in per disponibilità1 * 1 ST - SFr. 84.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Base for electronic crimping tool and high powered crimping toolBase for electronic crimping tool and high powered crimping toolSGEA5ABAS0Log in per disponibilità1 * 1 ST - SFr. 278.00