Orders LinkContactUsLinkComponent
CRS Battery-Powered Electronic Crimping Tools
Electronic decapper, 13 mm
Codice catalogo SGEA6A13D0
Nome del fornitore:   Trajan Scientific and Medical
CRS Battery-Powered Electronic Crimping Tools
Codice catalogo SGEA6A13D0
Nome del fornitore:  Trajan Scientific and Medical
CRS Battery-Powered Electronic Crimping Tools
CRS Battery-Powered Electronic Crimping Tools
Electronic decapper, 13 mm
Price
SFr. 1’130.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
The CRS battery-powered electronic crimping tools crimp/decap aluminum or two-part aluminum/steel caps with just the push of a button. The electronically adjustable cycle is accurate and reproducible to ensure consistently great results.
  • Powerful brushless motor with a fast cycle
  • Long lasting lithium ion battery provides up to 800 crimps/decaps per charge with an average recharge time of 1 to 2 hours
  • The tools are ergonomically designed to reduce strain associated with many manual crimping tools
  • The controls to increase or decrease crimp settlings are conveniently located on top of the unit, preventing accidental adjustment of the settings
  • OLED screen for simple operation displays tool size, battery status, crimp setting and cycle result
  • Performance tracking through an internal cycle log
  • Innovative charging system, which includes a warning light when recharging is necessary
  • Can be operated while charging
The names and trademarks of third party analytical instrument manufacturers are used solely to indicate the compatibility of Trajan products for use with such instruments.
Specifiche
  • Descrizione:
    Electronic decapper
  • Seq:
    08
  • Dimensione tappo:
    13 mm
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Stand for CRS electronic and hand crimpers and decappers
    Stand for CRS electronic and hand crimpers and decappers
    SGEA5ASTA0
    Log in per disponibilità
    1 * 1 ST - SFr. 84.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Base for electronic crimping tool and high powered crimping tool
    Base for electronic crimping tool and high powered crimping tool
    SGEA5ABAS0
    Log in per disponibilità
    1 * 1 ST - SFr. 278.00