PTFE GC Ferrules
GC ferrule 6,35 mm, no hole, PTFE, — mm
Codice catalogo SGEA2144XX
Nome del fornitore: Trajan Scientific and Medical
Vai a
Product Details & Documents
These GC ferrules are made from 100% PTFE. They are soft, completely inert, and have very low friction.
PTFE GC ferrules are hydrophobic and commonly used in liquid applications. They are rated upto upper temperature limit of 250 °C.
Specifiche
- Ø int.:— mm
- Descrizione:GC ferrule 6,35 mm, no hole
- Materiale:PTFE
- Seq:21
- Cat. No.:SGEA2144XX