PTFE GC Ferrules
GC ferrule 6,35 mm, PTFE, 6,0 mm
Codice catalogo SGEA214406
Nome del fornitore: Trajan Scientific and Medical
Vai a
Product Details & Documents
These GC ferrules are made from 100% PTFE. They are soft, completely inert, and have very low friction.
PTFE GC ferrules are hydrophobic and commonly used in liquid applications. They are rated upto upper temperature limit of 250 °C.
Specifiche
- Ø int.:6,0 mm
- Descrizione:GC ferrule 6,35 mm
- Materiale:PTFE
- Seq:18