Methyl 1-hydroxycyclohexanecarboxylate,95% 1 * 5 g
Catalog # AMBDA551192-5G
Supplier: AMBEED
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
ter-Butil metil etere, EMSURE® ACS per analisi, Supelco®
Supplier: Merck
Metilcicloesano per sintesi, Sigma-Aldrich®
Supplier: Merck
Etanolo assoluto ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Acetone ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Etere di petrolio, 60…95 °C, TECHNICAL
Supplier: VWR Chemicals
Acqua USP, JP, Ph. Eur., purificato
Supplier: VWR Chemicals
2-Propanolo ≥99.8%, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Metanolo ≥99.9%, HiPerSolv CHROMANORM®, ULTRA per LC-MS, per UPLC/UHPLC-MS
Supplier: VWR Chemicals
2-Propanolo ≥99.85%, CHROMASOLV™ per HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimetilformammide 99+%, extra puro
Supplier: Thermo Fisher Scientific
Etere etilico ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Formaldeide soluzione 35% (w/w) stabilizzato, meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Cicloesano 99+% per spettroscopia
Supplier: Thermo Fisher Scientific
2-Propanolo 99.5% per analisi
Supplier: Thermo Fisher Scientific
Etilen glicole ≥99%
Supplier: Thermo Fisher Scientific