5-Bromonaphthalen-1-amine,97% 1 * 25 g
Catalog # AMBDA216697-25G
Supplier: AMBEED
Frequently Bought Together
[EN]1,4-DIOXANE ANHYDROUS, 99.8 %, CONTA 1 * 1 L
Supplier: Merck
n-Pentano 99+%, extra puro
Supplier: Thermo Fisher Scientific
Diclorometano ≥99.9% (by GC) stabilizzato, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Etanolo denaturato ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Diclorometano 99.8% stabilizzato per HPLC
Supplier: Thermo Fisher Scientific
PETROLEUM ETHER 40-60C FOR PESTICIDE AN 1 * 2,5 L
Supplier: PanReac AppliChem
Cesio cloruro 99.99% (trace metals basis), Optical Grade per spettroscopia
Supplier: Thermo Fisher Scientific
ISOAMYL ALCOHOL ACCORDING TO GERBER FOR 1 * 1.000 mL
Supplier: PanReac AppliChem
Metanolo, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Diclorometano 99.9% stabilizzato, AcroSeal® per biochimica
Supplier: Thermo Fisher Scientific
Etanolo absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanolo, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
1,2-Diclorobenzene 99+% per HPLC
Supplier: Thermo Fisher Scientific