(1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazol-4-yl)boronic acid,96% 1 * 0,25 g
Catalog # AMBDA262727-250MG
Supplier: AMBEED
(1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazol-4-yl)boronic acid,96% 1 * 0,25 g
Catalog # AMBDA262727-250MG
Supplier: AMBEED
Frequently Bought Together
(±)-Alcol tetraidrofurfurilico, Sigma-Aldrich®
Supplier: Merck
Diclorometano stabilizzato, LiChrosolv® per HPLC, Supelco®
Supplier: Merck
Etere etilico ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanolo USP, J.T.Baker®
Supplier: Avantor
PETROLEUM ETHER 40-60C FOR PESTICIDE AN 1 * 2,5 L
Supplier: PanReac AppliChem
Cicloesano 99+% per spettroscopia
Supplier: Thermo Fisher Scientific
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
2-Metilbutano 99+%, extra puro
Supplier: Thermo Fisher Scientific
Esano (miscela di isomeri) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
Etanolo assoluto, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Nitrobenzene 99+% ACS
Supplier: Thermo Fisher Scientific