4-Fluoro-N-methylbenzamide,98% 1 * 0,25 g
Catalog # AMBDA868422-250MG
Supplier: AMBEED
Frequently Bought Together
n-Pentano ≥99.0%, AnalaR NORMAPUR® per analisi
Supplier: VWR Chemicals
1-Butanolo ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassio bromuro, Uvasol® per spettrofotometria IR, Supelco®
Supplier: Merck
Glicerina 87% 86%, AnalaR NORMAPUR® per analisi
Supplier: VWR Chemicals
Metanolo, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Acetone, LiChrosolv® per HPLC, Supelco®
Supplier: Merck
Cicloesano ≥99.5%, PESTINORM® per analisi dei pesticidi
Supplier: VWR Chemicals
Esano (miscela di isomeri), TECHNICAL
Supplier: VWR Chemicals
n-Eptano ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Glicerolo (glicerina) USP, Ph. Eur., BP
Supplier: VWR Chemicals
2-Propanolo ≥98%, TECHNICAL
Supplier: VWR Chemicals
Etanolo, GPR RECTAPUR®
Supplier: VWR Chemicals
Glicerina vegetale, anidro, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, JP per la produzione biofarmaceutica, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
n-Eptano 95%
Supplier: Thermo Fisher Scientific