Potassium trifluoro(4-methylnaphthalen-1-yl)borate,96% 1 * 0,25 g
Catalog # AMBDA318768-250MG
Supplier: AMBEED
Potassium trifluoro(4-methylnaphthalen-1-yl)borate,96% 1 * 0,25 g
Catalog # AMBDA318768-250MG
Supplier: AMBEED
Frequently Bought Together
2-Propanolo ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acqua, LiChrosolv® per cromatografia, per LC-MS, Supelco®
Supplier: Merck
N,N-Dimetilformammide ≥99.9%, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Acqua, TECHNICAL, demineralizzata
Supplier: VWR Chemicals
Glicerina ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Piridina ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
2-Propanolo ≥98%, TECHNICAL
Supplier: VWR Chemicals
2-Propanolo 70% ≥70% (v/v), GPR RECTAPUR®
Supplier: VWR Chemicals
Etanolo assoluto ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Etanolo assoluto ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Etanolo 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals