2-Methoxyphenothiazine,98% 1 * 5 g
AMBDA134702-5G
: AMBEED
Frequently Bought Together
Etilendiammina 99%
: Thermo Fisher Scientific
2-PROPANOL?á(USP, BP, PH. EUR.)?áPURE 1 * 200 L
: PanReac AppliChem
Dibutil-etere, extra dry 99+%, AcroSeal®
: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
: Thermo Fisher Scientific
Etile acetato, AR®, Laboratory Reagent, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
Tetrabutilammonio idrossido 0.4 M in water per HPLC
: Thermo Fisher Scientific
ter-Butil metil etere ≥99.0%, AnalaR NORMAPUR® ACS
: VWR Chemicals
2,2,4-Trimetil pentano 99.5% per analisi
: Thermo Fisher Scientific
ter-Butil metil etere, extra dry over Molecular Sieve 99+%, AcroSeal®
: Thermo Fisher Scientific
1,2-Diclorobenzene 99+% per HPLC
: Thermo Fisher Scientific