Orders LinkContactUsLinkComponent
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan apochromat, N.A. 100, 100×, 0,21 mm
Catalog # ECHLRMAG1013
Supplier:   DISCOVER ECHO
Objectives for Rebel Microscopes
Catalog # ECHLRMAG1013
Supplier:  DISCOVER ECHO
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan apochromat, N.A. 100, 100×, 0,21 mm
Price
SFr. 353.00
Log in for availability
1 * 1 ST
Specifications
  • Ingrandimento:
    100×
  • Descrizione:
    Plan apochromat, N.A. 100
  • Seq:
    07
  • Distanza di lavoro:
    0,21 mm