Orders LinkContactUsLinkComponent
Magnetic Stirring Bars
12 mm, 3 mm
  KART0076600
 :   KARTELL
Magnetic Stirring Bars
  KART0076600
 :  KARTELL
Magnetic Stirring Bars
12 mm, 3 mm
SFr. 40.30
1 * 10 ST
Economical stirring bars designed for a wide variety of applications.
  • Stronger mixing action at lower speeds
  • Can be used as an addition to any standard magnetic stir bar range, creating a viable alternative
  • The sizes shown below are nominal, with ±5% tolerance in length and ±10% in diameter
Suitable for foodstuff.
  • Ø:
    3 mm
  • Lunghezza:
    12 mm
  • Seq:
    01