Cole-Parmer® Essentials Replacement Cell Culturing Flasks
Glass cell culture flask only, 500 ml
TECN21837-32
Enhance the life of your BS-200 series cell culture stirring systems.
- Constructed of borosilicate glass
The culture vessels incorporate a unique base design which, together with the bulb-ended stirrer, ensures the cells are lifted into suspension at the lowest possible speeds. This gentle stirring action promotes high cell yields by preventing cell shearing. Replacement flasks do not include caps or stirring mechanisms.
- Capacità:500 ml
- Descrizione:Glass cell culture flask only
- Seq:03