(r)-(+)-n-benzyl-alpha-methylbenzylamine CAS: 38235-77-7 Catalog ID A252635 1*5g 1 * 5 g
AMBDA252635-5G
: AMBEED
(r)-(+)-n-benzyl-alpha-methylbenzylamine CAS: 38235-77-7 Catalog ID A252635 1*5g 1 * 5 g
AMBDA252635-5G
: AMBEED
Frequently Bought Together
Acetato di isopropile ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
: Honeywell Chemicals
PETROLEUM ETHER 40-60C FOR PESTICIDE AN 1 * 2,5 L
: PanReac AppliChem
Toluene ≥99.7%, CHROMASOLV™ for analysis of dioxins, furans and PCB, Riedel-de Haën™
: Honeywell Chemicals
1,2-Dicloroetano ≥99.8%, ChromAR® per HPLC, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
Carbonio solfuro 99.9% per HPLC
: Thermo Fisher Scientific
n-Pentano, anidro 99+%, extra puro
: Thermo Fisher Scientific
2-Propanolo ≥99.9%, CHROMASOLV™ Plus per HPLC, Riedel-de Haën™
: Honeywell Chemicals
n-Esano
: Roth Carl
N,N-Dimetilacetammide, extra dry 99.5%, AcroSeal®
: Thermo Fisher Scientific
Bromoformio 99+% stabilizzato
: Thermo Fisher Scientific
Acetone 99.6% ACS
: Thermo Fisher Scientific