Orders LinkContactUsLinkComponent
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan apochromat N.A. 0,40, 10×, 3,2 mm
Catalog # ECHLRMAG91022
Supplier:   DISCOVER ECHO
Objectives for Rebel Microscopes
Catalog # ECHLRMAG91022
Supplier:  DISCOVER ECHO
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan apochromat N.A. 0,40, 10×, 3,2 mm
Price
SFr. 1’520.00
Log in for availability
1 * 1 ST
Specifications
  • Ingrandimento:
    10×
  • Descrizione:
    Plan apochromat N.A. 0,40
  • Seq:
    19
  • Distanza di lavoro:
    3,2 mm