CPC® Miniature Quick-Disconnect Fitting, Panel-Mount Hose Barb Insert, Acetal, Straight-Through, 3 mm ID, SMMPMM3; 50/PK
Codice catalogo MFLX06363-71BLK
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
No more kinked tubing—allows tubing to rotate freely.
- Twist-to-connect couplings provide a reliable and more secure alternative to luer-type connections
- Tubing can rotate freely when connected—preventing kinked tubing and accidental disconnection
These CPC® miniature quick-disconnect couplings are able to handle 1/16" ID tubing, and offer a more secure connection than luer fittings. Fitting halves engage and seal with a twist. Once engaged, the coupling allows the tubing to rotate freely.
For a complete coupling, order one fitting body and one insert.
- Dimensioni del raccordo/Ø int. del tubo (porta 1):3 mm
- Tipo di connessione:Quick-Disconnect to Barb
- Flow pattern:Straight
- Temperatura massima:82 °C (180 °F)
- Descrizione:Miniature Quick-Disconnect Fitting, Panel-Mount Hose Barb Insert, Acetal, Straight-Through, 3 mm ID, SMMPMM3; 50/PK
- Environmentally Preferable:
- O-Ring material:Buna N
- Valve material:None
- Temperatura minima:–40 °C (–40 °F)
- Sterilizzazione:Autoclave or ethylene oxide (ETO)
- Garanzia:1 year
- Conf:50
- Fitting material:Acetal
- Quick-disconnect flow size:³⁄₃₂"
- Supplier No.:SMMPMM3 50PK
Frequently Bought Together