L(+)-Serina USP
12 kg
SPCMS1023-12KGBL
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine
- :56-45-1
- :C₃H₇NO₃
- :MFCD00064224
- :105.09 g/mol
- :14,00846
- :259 °C (1013 hPa)
- :215…225 °C
- :1,6 g/cm³ (22 °C)
- :Ambiente
- :200-274-3
Documents
- Assay:98.5% Min.
- Conf:12 kg