Additional shelves for FD Range Ovens
Stainless steel shelf, for heavy loads, max load 70 kg, Reels BF/FD/FED 720
Codice catalogo BIND8012-2185
Nome del fornitore: Binder
Vai a
Product Details & Documents
Specifiche
- Descrizione:Stainless steel shelf, for heavy loads, max load 70 kg
- Seq:05
- Per:Reels BF/FD/FED 720