MOUNTAIN PRI OPTOGENETICS FIBER 500-1.25 HIGH NA POLYMER OPTICAL FIBER CONNECTING THE ROTARY JOINT TO THE IMPLANTED 1.25 MM DIAMETER CANNULA FC CONNECTOR ON ONE END AND 1.25 MM FERRULE ON OTHER END FIBER CORE 500 UM NA 0.63 FIBER LENGTH 1 M 1 * 1 m
Codice catalogo MSPP34122
Nome del fornitore: MARKET SOURCE PART PROCESS
Frequently Bought Together