Orders LinkContactUsLinkComponent
Wax, Reference Standards
Wax, Reference Standards
White Wax, USP reference standard, Vial, 30 mg
Codice catalogo 1719218.
Nome del fornitore:   USP
Wax, Reference Standards
Codice catalogo 1719218.
Nome del fornitore:  USP
Numero CAS:  
Wax, Reference Standards
White Wax, USP reference standard, Vial, 30 mg
Numero CAS:  
Price
SFr. 914.00
Log in per disponibilità
1 * 30 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Zertifizierung: US Pharmacopea Reference Standard
Specifiche
Specifiche
  • Tipo di conf.:
    Vial
  • Conf:
    30 mg
  • Standard:
    USP reference standard
  • Descrizione:
    White Wax
  • Cat. No.:
    1719218.
  • Descrizione:
    White Wax
  • CAS No.:
    8012-89-3
Specifiche
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..