Human IL-17F ELISpot Kit
Human IL-17F ELISpot kit
Catalog # ABCAAB281968-1X96
Supplier: Abcam
Jump to:
Product Details & Documents
Human IL-17F ELISpot Kit is a ELISpot Kit for the sensitive detection of Human IL-17F production in a single cell suspension.
Specifications
Specifications
- Descrizione:Human IL-17F ELISpot kit
- Cat. No.:ABCAAB281968-1X96
- Forma:Pre-coated
- Simbolo dell'antigene:IL-17F
- Stato regolamentazione:RUO
- Tiipo del campione:Suspension cells
- Temperatura di conservazione:4 °C
- Tempo di elaborazione:Multiple steps
- Reattività delle specie:Human
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



