Masterflex® Single-Use Bag Port Disc Mini Dome, Avantor®
Port disc ¾" barb in engage 8402 cleanroom manufactured; 10 pack, ¾", Port Disc to Barb, Engage 8402, Straight
Codice catalogo MFLX40331-01
Nome del fornitore: Avantor Fluid Handling
Masterflex® Single-Use Bag Port Disc Mini Dome, Avantor®
Codice catalogo MFLX40331-01
Nome del fornitore: Avantor Fluid Handling
Vai a
Product Details & Documents
Mini-dome protrusions inhibit suction blockage.
- Barb sizes range from 1/8" to 1"
- 2-1/4" disc diameter
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets.
If material validation is needed, please reach out to our techincal support representatives.
Specifiche
- Dimensioni del raccordo/Ø int. del tubo (porta 1):¾"
- Tipo di connessione:Port Disc to Barb
- Flow pattern:Straight
- Descrizione:Port disc ¾" barb in engage 8402 cleanroom manufactured; 10 pack
- Fitting material:Engage 8402
- Cat. No.:MFLX40331-01
- Regulatory approvals:FDA;USP
- Sterilizzazione:Gamma irradiation