Masterflex® Cleanroom 1" Sanitary Flange × ¾" Hose Barb in Polypropylene; 10 pack
Catalog # MFLX40330-24
Supplier: Avantor Fluid Handling
Specifications
See all product configurations- Catalog No:
- MFLX40330-24
- MFLX40330-35
- MFLX40330-25
- MFLX40330-36
- MFLX40330-26
- MFLX40330-37
- MFLX40330-27
- MFLX40330-38
- MFLX40330-32
- MFLX40330-22
- MFLX40330-33
- MFLX40330-23
- MFLX40330-34
- MFLX40330-28
- MFLX40330-39
- MFLX40330-29
- MFLX40330-40
- MFLX40330-30
- MFLX40330-41
- MFLX40330-31
- No. of ports:
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- Dimensione flangia:
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- 1"
- Sterilizzazione:
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Gamma irradiation, EtO, autoclave
- Regulatory approvals:
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- FDA;USP
- Currently Viewing
Specifications
About this item
Maintain sanitary conditions with secure connections from sanitary Tri-Clamp to tubing.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Packaged to exacting tolerances under ISO Class 7 cleanroom conditions
- Compatible with other Tri-Clamp® connections
The sanitary Tri-Clamp barbed adapters are ideal for use in the biopharmaceutical, food and beverage, and single-use industries. They have excellent chemical resistance and may be sterilized by gamma irradiation, EtO, E-beam, or autoclave. The PVDF fittings withstand higher temperatures than the Polypropylene fittings. Clamps and gaskets required for a complete system are sold separately.
- Dimensioni del raccordo/Ø int. del tubo (porta 2):¾"
- Tipo di connessione:Sanitary to Barb
- Flow pattern:Straight
- Sanitary clamp size (Port 1):1½"
- Descrizione:Cleanroom 1½" sanitary tri-clamp × ¾" barb adapter in polypropylene; 10 pack
- Seq:14
- Environmentally Preferable:
- No. of ports:2
- Regulatory approvals:FDA;USP
- Sterilizzazione:Gamma irradiation, EtO, autoclave
- Dimensione flangia:1"
- Conf:10
- Fitting material:Polypropylene
- Sanitary gasket size:1½"
Frequently Bought Together