Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
[EN]HELMET V-GARD 950 NON VENT FAS-TRAC 1 * 12 pezzi
MSAAGVF1AC0A00H000
:
M S A
[EN]HELMET V-GARD 950 NON VENT FAS-TRAC 1 * 12 pezzi
MSAAGVF1AC0A00H000
:
M S A
Frequently Bought Together
Guanti in nitrile, TouchNTuff® 92-600 / 92-605
: Ansell
Guanti in nitrile, Kimtech™ Purple Nitrile™ Xtra™
: KIMBERLY CLARK
VWR®, Etichette di sicurezza, secondo il sistema GHS
: VWR Collection
VWR® NITRILE LIGHT, Guanti in nitrile
: VWR Collection
Guanti in nitrile, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Colour Lab Tapes
: GA
Shoe covers, CPE
: HUBEI ORIENT INTL TRADING
VWR® Tute per protezione da prodotti chimici
: VWR Collection
Shoe covers, PP/CPE
: HUBEI ORIENT INTL TRADING
Overalls, 4500
: 3M
VWR®, Guanti in lattice
: VWR Collection