Masterflex® L/S® Easy-Load® Pump Head Replacement Rotor/Bearing Assembly 07514-12 and -32
Codice catalogo MFLX07514-19
Nome del fornitore: Avantor Fluid Handling
1 year warranty
Informazioni su questo articolo
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:L/S
- Descrizione:L/S® Easy-Load® Pump Head Replacement Rotor/Bearing Assembly 07514-12 and -32
- Environmentally Preferable:
- Open-head sensor:No
- Tubing sizes accepted:L/S® 15, L/S 24
- Garanzia:1 year
- Conf:1
- Rulli:4
- Rotor material:Stainless Steel
Frequently Bought Together