ContactUsLinkComponent
FITC-Labeled Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 is expressed from human HEK293 cells. It is the FITC labeled form of Mo noclonal Anti-FMC63 scFv Antibody, Mouse IgG1 (Cat. No. FM3-Y45P1). 1 * 25 Tests
Catalog # ACRBFM3-FY45P1-25
Supplier:  ACRO Biosystems
FITC-Labeled Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 is expressed from human HEK293 cells. It is the FITC labeled form of Mo noclonal Anti-FMC63 scFv Antibody, Mouse IgG1 (Cat. No. FM3-Y45P1). 1 * 25 Tests
Catalog # ACRBFM3-FY45P1-25
Supplier:  ACRO Biosystems
Frequently Bought Together