ContactUsLinkComponent
Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 is isolated from a SARS-CoV-2 infected patient and is recombinantly produced from CHO cells. This monoclonal antibody is purified by Protein A affinity chromatography. As verified by binding test with 1 * 200 µG
  ACRBNUN-CH14-200UG
Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 is isolated from a SARS-CoV-2 infected patient and is recombinantly produced from CHO cells. This monoclonal antibody is purified by Protein A affinity chromatography. As verified by binding test with 1 * 200 µG
  ACRBNUN-CH14-200UG
Frequently Bought Together