ContactUsLinkComponent
Human Siglec-10, Fc Tag (SI0-H5254) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Accession # Q96LC7- 1). 1 * 1 mg
  ACRBSI0-H5254-1MG
Human Siglec-10, Fc Tag (SI0-H5254) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Accession # Q96LC7- 1). 1 * 1 mg
  ACRBSI0-H5254-1MG
Frequently Bought Together