ContactUsLinkComponent
FITC-Labeled Human EGF R, Fc Tag (EGR-HF256) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1). It is the FITC labeled form of Human EGF R, Fc Tag (Cat. No. EGR-H5252). 1 * 200 µG
Catalog # ACRBEGR-HF256200UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human EGF R, Fc Tag (EGR-HF256) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1). It is the FITC labeled form of Human EGF R, Fc Tag (Cat. No. EGR-H5252). 1 * 200 µG
Catalog # ACRBEGR-HF256200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together