FITC-Labeled Human GUCY2C, Fc Tag (GUC-HF255) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # P25092-1). 1 * 25 µG
Codice catalogo ACRBGUC-HF255-25UG
Nome del fornitore: ACROBIOSYSTEMS
Frequently Bought Together