Human SSTR2 Full Length Protein-VLP (SS2-H5216) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ile 369 (Accessio n # P30874-1). 1 * 20 µG
Human SSTR2 Full Length Protein-VLP (SS2-H5216) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ile 369 (Accessio n # P30874-1). 1 * 20 µG