ContactUsLinkComponent
Biotinylated Human GUCY2D, His,Avitag (GUD-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ala 52 - Glu 462 (Acces sion # Q02846-1). 1 * 25 µG
Codice catalogo ACRBGUD-H82E6-25UG
Nome del fornitore:  ACRO Biosystems
Biotinylated Human GUCY2D, His,Avitag (GUD-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ala 52 - Glu 462 (Acces sion # Q02846-1). 1 * 25 µG
Codice catalogo ACRBGUD-H82E6-25UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together