ContactUsLinkComponent
Biotinylated Human APOE4 His,Avitag (APE-H52E7) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Accessi on # P02649-1 (C130R)). 1 * 200 µG
Catalog # ACRBAPE-H52E7200UG
Supplier:  ACRO Biosystems
Biotinylated Human APOE4 His,Avitag (APE-H52E7) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Accessi on # P02649-1 (C130R)). 1 * 200 µG
Catalog # ACRBAPE-H52E7200UG
Supplier:  ACRO Biosystems
Frequently Bought Together