ContactUsLinkComponent
Human NKG2A&CD94, Fc Tag (NC4-H5257) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Leu 233 (NKG2A) & Asp 57 - Ile 179 (CD94) (Accession # P26715-1 (NKG2A) & Q13241-1 (CD94)). 1 * 1 mg
Codice catalogo ACRBNC4-H5257-1MG
Nome del fornitore:  ACRO Biosystems
Human NKG2A&CD94, Fc Tag (NC4-H5257) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Leu 233 (NKG2A) & Asp 57 - Ile 179 (CD94) (Accession # P26715-1 (NKG2A) & Q13241-1 (CD94)). 1 * 1 mg
Codice catalogo ACRBNC4-H5257-1MG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together