ContactUsLinkComponent
Canine CD19, His Tag (CD9-C52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Gly 297 (Accession # XP_0385246 89.1). 1 * 1 mg
Codice catalogo ACRBCD9-C52H4-1MG
Nome del fornitore:  ACRO Biosystems
Canine CD19, His Tag (CD9-C52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Gly 297 (Accession # XP_0385246 89.1). 1 * 1 mg
Codice catalogo ACRBCD9-C52H4-1MG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together