ContactUsLinkComponent
FITC-Labeled Human Flt-3, Fc Tag (FL3-HF255) is expressed from human 293 cells (HEK293). It contains AA Asn 27 - Ser 543 (Accession # P36888-1). 1 * 25 µG
Codice catalogo ACRBFL3-HF255-25UG
Nome del fornitore:  ACROBIOSYSTEMS
FITC-Labeled Human Flt-3, Fc Tag (FL3-HF255) is expressed from human 293 cells (HEK293). It contains AA Asn 27 - Ser 543 (Accession # P36888-1). 1 * 25 µG
Codice catalogo ACRBFL3-HF255-25UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together