ContactUsLinkComponent
FITC-Labeled Human HGF R, His Tag (HGR-HF224) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Thr 932 (Accession # AAI30421.1). 1 * 200 µG
Codice catalogo ACRBHGR-HF224200UG
Nome del fornitore:  ACROBIOSYSTEMS
FITC-Labeled Human HGF R, His Tag (HGR-HF224) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Thr 932 (Accession # AAI30421.1). 1 * 200 µG
Codice catalogo ACRBHGR-HF224200UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together