ContactUsLinkComponent
FITC-Labeled Human Carbonic Anhydrase IX (38-414), His Tag (CA9-HF228) is expressed from human 293 cells (HEK293). It contains AA Gl n 38 - Asp 414 (Accession # Q16790-1). 1 * 25 µG
Codice catalogo ACRBCA9-HF228-25UG
Nome del fornitore:  ACROBIOSYSTEMS
FITC-Labeled Human Carbonic Anhydrase IX (38-414), His Tag (CA9-HF228) is expressed from human 293 cells (HEK293). It contains AA Gl n 38 - Asp 414 (Accession # Q16790-1). 1 * 25 µG
Codice catalogo ACRBCA9-HF228-25UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together