ContactUsLinkComponent
GMP Human IL-7 (GMP-L07H24) is expressed from human 293 cells (HEK293). It contains AA Asp 26 - His 177 (Accession # P13232-1). 1 * 50 µG
Codice catalogo ACRBGMP-L07H2450UG
Nome del fornitore:  ACRO Biosystems
GMP Human IL-7 (GMP-L07H24) is expressed from human 293 cells (HEK293). It contains AA Asp 26 - His 177 (Accession # P13232-1). 1 * 50 µG
Codice catalogo ACRBGMP-L07H2450UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together