Biotinylated Human LAG-3, His,Avitag (LA3-H82E9) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Access ion # P18627-1). 1 * 200 µG
Codice catalogo ACRBLA3-H82E9200UG
Nome del fornitore: ACRO Biosystems
Frequently Bought Together