ContactUsLinkComponent
Human CLEC-2, Fc Tag (CL2-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 55 - Pro 229 (Accession # Q9P126-1). 1 * 100 µG
  ACRBCL2-H5254100UG
Human CLEC-2, Fc Tag (CL2-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 55 - Pro 229 (Accession # Q9P126-1). 1 * 100 µG
  ACRBCL2-H5254100UG
Frequently Bought Together