ContactUsLinkComponent
Biotinylated Human HLA-A*1101&B2M Complex Protein (HLM-H82W5) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Th r 305 (HLA-G) & Ile 21 - Met 119 (B2M) (Accession # Q5S3G3-1 (HLA-G) & P61769-1 (B2M)). 1 * 25 µG
Codice catalogo ACRBHLM-H82W5-25UG
Nome del fornitore:  ACRO Biosystems
Biotinylated Human HLA-A*1101&B2M Complex Protein (HLM-H82W5) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Th r 305 (HLA-G) & Ile 21 - Met 119 (B2M) (Accession # Q5S3G3-1 (HLA-G) & P61769-1 (B2M)). 1 * 25 µG
Codice catalogo ACRBHLM-H82W5-25UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together