ContactUsLinkComponent
Rabbit TROP-2, His Tag (TR2-R52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Pro 279 (Accession # G1T6B6-1 ). 1 * 100 µG
Codice catalogo ACRBTR2-R52H9100UG
Nome del fornitore:  ACRO Biosystems
Rabbit TROP-2, His Tag (TR2-R52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Pro 279 (Accession # G1T6B6-1 ). 1 * 100 µG
Codice catalogo ACRBTR2-R52H9100UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together