ContactUsLinkComponent
FITC-Labeled Human CD99, Fc Tag (CD9-HF254) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 122 (Accession # P14209-1). 1 * 200 µG
Catalog # ACRBCD9-HF254200UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human CD99, Fc Tag (CD9-HF254) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 122 (Accession # P14209-1). 1 * 200 µG
Catalog # ACRBCD9-HF254200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together