ContactUsLinkComponent
Monkeypox virus (strain Zaire-96-I-16) A30L, His Tag (A3L-M5243) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Leu 146 (Accession # Q8V4U9-1). 1 * 500 µG
Codice catalogo ACRBA3L-M5243500UG
Nome del fornitore:  ACROBIOSYSTEMS
Monkeypox virus (strain Zaire-96-I-16) A30L, His Tag (A3L-M5243) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Leu 146 (Accession # Q8V4U9-1). 1 * 500 µG
Codice catalogo ACRBA3L-M5243500UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together